• USA Home
  • Search Results

  • aziprotryne22527465828011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Aziprotryne

1 Products
Synonym:   2-Azido-4-isopropylamino-6-methylthio-1,3,5-triazine
Image of Aziprotryne
CAS Number:
Empirical Formula (Hill Notation):
 C7H11N7S
Molecular Weight:
 225.27
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
Show More