• USA Home
  • Search Results

  • azithromycindihydrate7850211777270011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Azithromycin dihydrate

1 Products
Synonym:   N-Methyl-11-aza-10-deoxo-10-dihydroerythromycin A, CP-62993
Image of Azithromycin dihydrate
CAS Number:
Empirical Formula (Hill Notation):
 C38H72N2O12 · 2H2O
Molecular Weight:
 785.02
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More