• USA Home
  • Search Results

  • azoxystrobin4033913186033811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Azoxystrobin

2 Products
Image of Azoxystrobin
CAS Number:
Empirical Formula (Hill Notation):
 C22H17N3O5
Molecular Weight:
 403.39
 Colour Index Number:
 23860
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Shelf Life
  Special Grade
Show More