• USA Home
  • Search Results

  • azureachloride2918053153311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Azure A chloride

3 Products
Image of Azure A chloride
CAS Number:
Empirical Formula (Hill Notation):
 C14H14ClN3S
Molecular Weight:
 291.80
 Beilstein/REAXYS Number:
 3922287
 Colour Index Number:
 52005
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Chemical Composition
  Special Grade
Show More