• USA Home
  • Search Results

  • bacetylgoalkyllaphosphatidylcholinefrombovineheartlecithin123456515406511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

β-Acetyl-γ-O-alkyl-L-α-phosphatidylcholine from bovine heart lecithin

1 Products
Synonym:   1-O-Alkyl-2-acetyl-sn-glycero-3-phosphocholine, 2-Acetyl-1-alkyl-sn-glycero-3-phosphocholine, AcGEPC, PAF, Platelet activating factor
Image of β-Acetyl-γ-O-alkyl-<SC>L</SC>-α-phosphatidylcholine from bovine heart lecithin
CAS Number:
 Beilstein/REAXYS Number:
 4220745
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  Product Information Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More