• USA Home
  • Search Results

  • bacitracinzinc148607140589611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Bacitracin zinc

2 Products
Synonym:   Bacitracin zinc salt, Zinc bacitracin
Image of Bacitracin zinc
CAS Number:
Empirical Formula (Hill Notation):
 C66H101N17O16SZn
Molecular Weight:
 1486.07
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Physical Form
  Application(s)
  Anal. Standard Form
Show More