• USA Home
  • Search Results

  • bacitracinzincsalt148607140589611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Bacitracin zinc salt

2 Products
Synonym:   Zinc bacitracin
Image of Bacitracin zinc salt
CAS Number:
Empirical Formula (Hill Notation):
 C66H101N17O16SZn
Molecular Weight:
 1486.07
 EC Number:
 MDL number:
 NACRES:
 NA.85
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Biological Source
  Mode of Action
  Activity Spectrum
  Physical Form
Show More