• USA Home
  • Search Results

  • bacteriologicalagar12345900218011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Bacteriological agar

1 Products
Synonym:   Agar-agar, Agar, Gum agar
CAS Number:
Linear Formula:
 (C12H18O9)n
 EC Number:
 MDL number:
 NACRES:
 NA.31
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More