• USA Home
  • Search Results

  • baicalein7diglucoside5945211448286911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Baicalein 7-diglucoside

1 Products
Synonym:   5,6,7-Trihydroxyflavone 7-diglucoside, Baicalein 7-gentiobioside, Baicalein-7-O-diglucoside, Oroxin B
Image of Baicalein 7-diglucoside
CAS Number:
Empirical Formula (Hill Notation):
 C27H30O15
Molecular Weight:
 594.52
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More