• USA Home
  • Search Results

  • balanine890910795911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

β-Alanine

6 Products
Synonym:   β-Ala, 3-Aminopropionic acid
Image of β-Alanine
CAS Number:
Linear Formula:
 NH2CH2CH2COOH
Molecular Weight:
 89.09
 Beilstein/REAXYS Number:
 906793
 EC Number:
 MDL number:
 NACRES:
 NA.26
 PubChem Substance ID:
 eCl@ss:
 39030936
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More