• USA Home
  • Search Results

  • bamanitinfromamanitaphalloides919952115022111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

β-Amanitin from Amanita phalloides

1 Products
Image of β-Amanitin from <I>Amanita phalloides</I>
CAS Number:
Empirical Formula (Hill Notation):
 C39H53N9O15S
Molecular Weight:
 919.95
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More