• USA Home
  • Search Results

  • barbital184195744311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Barbital

3 Products
Synonym:   5,5-Diethyl-2,4,6(1H,3H,5H)-pyrimidinetrione, 5,5-Diethylbarbituric acid, Barbitone, Veronal
Image of Barbital
CAS Number:
Empirical Formula (Hill Notation):
 C8H12N2O3
Molecular Weight:
 184.19
 Beilstein/REAXYS Number:
 163999
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
  Application(s)
Show More