• USA Home
  • Search Results

  • barbitalsolution184195744311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Barbital solution

1 Products
Image of Barbital solution
CAS Number:
Empirical Formula (Hill Notation):
 C8H12N2O3
Molecular Weight:
 184.19
 MDL number:
 NACRES
 NA.24
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More