• USA Home
  • Search Results

  • barium13733744039311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Barium

5 Products
Image of Barium
CAS Number:
Empirical Formula (Hill Notation):
 Ba
Molecular Weight:
 137.33
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Purification method
  Particle Dimension
  Physical Form
Show More