• USA Home
  • Search Results

  • bariumacetate2554254380611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Barium acetate

4 Products
Image of Barium acetate
CAS Number:
Linear Formula:
 (CH3COO)2Ba
Molecular Weight:
 255.42
 Beilstein/REAXYS Number:
 3693411
 EC Number:
 MDL number:
 NACRES:
 NB.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  pH Value
  Special Grade
  Physical Form
Show More