• USA Home
  • Search Results

  • bariumcarbonate1973451377911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Barium carbonate

4 Products
Synonym:   Barium carbonate
Image of Barium carbonate
CAS Number:
Empirical Formula (Hill Notation):
 CBaO3
Molecular Weight:
 197.34
 EC Index Number:
 208-167-3
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Physical Form
  Restriction
  Purity Grade
  Special Grade
Show More