• USA Home
  • Search Results

  • bariumchloride208231036137211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Barium chloride

5 Products
Synonym:   Barium dichloride, Barium dichloride
Image of Barium chloride
CAS Number:
Empirical Formula (Hill Notation):
 BaCl2
Molecular Weight:
 208.23
 EC Index Number
 056-004-00-8
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
  Particle Dimension
  Physical Form
Show More