• USA Home
  • Search Results

  • bariumchromate253321029440311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Barium chromate

2 Products
Synonym:   Barium chromium oxide (BaCrO4 ), Baryta yellow
Image of Barium chromate
CAS Number:
Empirical Formula (Hill Notation):
 BaCrO4
Molecular Weight:
 253.32
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Physical Form
Show More