• USA Home
  • Search Results

  • bariumhydroxideoctahydrate315461223071611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Barium hydroxide octahydrate

4 Products
Image of Barium hydroxide octahydrate
CAS Number:
Linear Formula:
 Ba(OH)2 · 8H2O
Molecular Weight:
 315.46
 EC Number:
 MDL number:
 NACRES:
 NA.55
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Restriction
  Potency
  Melting Point
  Density
  Bulk Density
  Agency/Method
  Special Grade
  Physical Form
  pH Value
Show More