• USA Home
  • Search Results

  • bariumiodidedihydrate42717778733911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Barium iodide dihydrate

2 Products
Image of Barium iodide dihydrate
CAS Number:
Linear Formula:
 Bal2 · 2H2O
Molecular Weight:
 427.17
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More