• USA Home
  • Search Results

  • bariumtitanateiv233191204727711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Barium titanate (IV)

7 Products
Synonym:   Barium metatitanate, Barium titanate (IV)
Image of Barium titanate (IV)
CAS Number:
Empirical Formula (Hill Notation):
 BaO3Ti
Molecular Weight:
 233.19
 EC Index Number
 234-975-0
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  Reaction Suitability
  Particle Dimension
  Assay
Show More