• USA Home
  • Search Results

  • bavachinin338401987930211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Bavachinin

2 Products
Synonym:   4′-Hydroxy-7-methoxy-6-(3-methyl-2-butenyl)flavanone, 7-O-Methylbavachin, Bavachinin A
Image of Bavachinin
CAS Number:
Empirical Formula (Hill Notation):
 C21H22O4
Molecular Weight:
 338.40
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More