• USA Home
  • Search Results

  • bcarboline3carboxylicacidnmethylamide225257853874611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

β-Carboline-3-carboxylic acid N-methylamide

1 Products
Synonym:   β-CCA methylamide, FG-7142, N-Methyl-β-carboline-3-carboxamide
Image of β-Carboline-3-carboxylic acid N-methylamide
CAS Number:
Empirical Formula (Hill Notation):
 C13H11N3O
Molecular Weight:
 225.25
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More