• USA Home
  • Search Results

  • bchlorolalaninehydrochloride160005188789911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

β-Chloro-L-alanine hydrochloride

1 Products
Image of β-Chloro-<SC>L</SC>-alanine hydrochloride
CAS Number:
Empirical Formula (Hill Notation):
 C3H6ClNO2 · HCl
Molecular Weight:
 160.00
 MDL number:
 NACRES:
 NA.32
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More