• USA Home
  • Search Results

  • bcitronellol1562710622911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(±)-β-Citronellol

4 Products
Synonym:   3,7-Dimethyl-6-octen-1-ol
Image of (±)-β-Citronellol
CAS Number:
Linear Formula:
 (CH3)2C=CHCH2CH2CH(CH3)CH2CH2OH
Molecular Weight:
 156.27
 Beilstein/REAXYS Number:
 1721507
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More