• USA Home
  • Search Results

  • bcyclodextrin113498758539911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

β-Cyclodextrin

4 Products
Synonym:   Caraway, Cycloheptaamylose, Cyclomaltoheptaose, Schardinger β-Dextrin, beta-Cyclodextrin
Image of β-Cyclodextrin
CAS Number:
Empirical Formula (Hill Notation):
 C42H70O35
Molecular Weight:
 1134.98
 Beilstein/REAXYS Number:
 78623
 EC Number:
 MDL number:
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More