• USA Home
  • Search Results

  • bcyclodextrinhydrate113498anhydrousbasis6816823011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

β-Cyclodextrin hydrate

1 Products
Synonym:   β-Schardinger dextrin, Cycloheptaamylose, Schardinger β-Dextrin hydrate, beta-Cyclodextrin hydrate
Image of β-Cyclodextrin hydrate
CAS Number:
Empirical Formula (Hill Notation):
 C42H70O35 · xH2O
Molecular Weight:
 1134.98 (anhydrous basis)
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More