• USA Home
  • Search Results

  • beauvericin783952604805511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Beauvericin

2 Products
Synonym:   cyclo(D-α-Hydroxyisovaleryl-L-N-methyl-Phe)3
Image of Beauvericin
CAS Number:
Empirical Formula (Hill Notation):
 C45H57N3O9
Molecular Weight:
 783.95
 MDL number:
 NACRES:
 NA.26
 PubChem Substance ID:
Popular Documents:  Product Information Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Physical Form
Show More