• USA Home
  • Search Results

  • beclomethasone40892441939011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Beclomethasone

1 Products
Synonym:   9α-Chloro-11β,17α,21-trihydroxy-16β-methylpregna-1,4-diene-3,20-dione, 9α-Chloro-16β-methyl-1,4-pregnadiene-11β,17α,21-triol-3,20-dione
Image of Beclomethasone
CAS Number:
Empirical Formula (Hill Notation):
 C22H29ClO5
Molecular Weight:
 408.92
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
B0385 Beclomethasone, ≥99%,