• USA Home
  • Search Results

  • beclomethasonedipropionate52104553409811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Beclomethasone dipropionate

4 Products
Synonym:   9-Chloro-11β,17,21-trihydroxy-16β-methylpregna-1,4-diene-3,20-dione 17,21-dipropionate
Image of Beclomethasone dipropionate
CAS Number:
Empirical Formula (Hill Notation):
 C28H37ClO7
Molecular Weight:
 521.04
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Titertest
  Anal. Standard Form
  Physical Form
Show More