• USA Home
  • Search Results

  • beflubutamid3553311361408711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Beflubutamid

1 Products
Synonym:   2-[4-Fluoro-3-(trifluoromethyl)phenoxy]-N-(phenylmethyl)butanamide, N-Benzyl-2-(α,α,α,4-tetrafluoro-m-tolyloxy)butyramide
Image of Beflubutamid
CAS Number:
Empirical Formula (Hill Notation):
 C18H17F4NO2
Molecular Weight:
 355.33
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
Show More