• USA Home
  • Search Results

  • behenicacid3405811285611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Behenic acid

2 Products
Synonym:   Docosanoic acid
Image of Behenic acid
CAS Number:
Linear Formula:
 CH3(CH2)20COOH
Molecular Weight:
 340.58
 Beilstein/REAXYS Number:
 1792887
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Application(s)
  Solubility
Show More