• USA Home
  • Search Results

  • benactyzinehydrochloride363885737411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Benactyzine hydrochloride

1 Products
Synonym:   α-Hydroxy-α-phenyl-benzeneacetic acid 2-(diethylamino)ethyl ester, α-Hydroxy-α-phenylbenzeneacetic acid 2-(diethylamino)ethyl ester, 2-(Diethylamino)ethyl benzilate hydrochloride, 2-Diethylaminoethyl benzilate, Benactyzine, Benzilic acid 2-(diethylamino)ethyl ester
Image of Benactyzine hydrochloride
CAS Number:
Linear Formula:
 (C6H5)2C(OH)CO2CH2CH2N(C2H5)2·HCl
Molecular Weight:
 363.88
 EC Number:
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More