• USA Home
  • Search Results

  • benazeprilhydrochloride460958654174411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Benazepril hydrochloride

4 Products
Synonym:   (3S)-3-[[(1S)-1-(Ethoxycarbonyl)-3-phenylpropyl]amino]-2,3,4,5-tetrahydro-2-oxo-1H-1-benzazepine-1-acetic acid hydrochloride, Benazepril hydrochloride
Image of Benazepril hydrochloride
CAS Number:
Empirical Formula (Hill Notation):
 C24H28N2O5 · HCl
Molecular Weight:
 460.95
 MDL number:
 NACRES
 NA.24
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Physical Form
  Manufacturer designation
Show More