• USA Home
  • Search Results

  • benazolinethylester271722505980711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Benazolin-ethyl ester

1 Products
Synonym:   4-Chloro-2-oxo-3(2H)-benzothiazoleacetic acid ethyl ester, Ethyl-4-chlor-2-oxo-benzothiazolinacetat
Image of Benazolin-ethyl ester
CAS Number:
Empirical Formula (Hill Notation):
 C11H10ClNO3S
Molecular Weight:
 271.72
 Beilstein/REAXYS Number:
 753659
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
Show More