• USA Home
  • Search Results

  • benfuresate256326850569111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Benfuresate

1 Products
Image of Benfuresate
CAS Number:
Empirical Formula (Hill Notation):
 C12H16O4S
Molecular Weight:
 256.32
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
Show More