• USA Home
  • Search Results

  • benodanil323131531001711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Benodanil

1 Products
Synonym:   2-Iodobenzanilide
Image of Benodanil
CAS Number:
Empirical Formula (Hill Notation):
 C13H10INO
Molecular Weight:
 323.13
 Beilstein/REAXYS Number:
 2725018
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
Show More