• USA Home
  • Search Results

  • benomyl290321780435211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Benomyl

1 Products
Synonym:   Benomyl, Methyl 1-(butylcarbamoyl)-2-benzimidazolecarbamate, NSC 263489
Image of Benomyl
CAS Number:
Empirical Formula (Hill Notation):
 C14H18N4O3
Molecular Weight:
 290.32
 Beilstein/REAXYS Number:
 825455
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Anal. Standard Form
  Physical Form
  Special Grade
Show More