• USA Home
  • Search Results

  • benoxacor260129873004211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Benoxacor

2 Products
Image of Benoxacor
CAS Number:
Empirical Formula (Hill Notation):
 C11H11Cl2NO2
Molecular Weight:
 260.12
 Beilstein/REAXYS Number:
 4190275
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Shelf Life
  Special Grade
Show More