• USA Home
  • Search Results

  • benserazidehydrochloride293701491977811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Benserazide hydrochloride

3 Products
Synonym:   DL-Serine 2-(2,3,4-trihydroxybenzyl)hydrazide hydrochloride
Image of Benserazide hydrochloride
CAS Number:
Empirical Formula (Hill Notation):
 C10H15N3O5 · HCl
Molecular Weight:
 293.70
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Manufacturer designation
  Physical Form
  Special Grade
Show More