• USA Home
  • Search Results

  • bensulide3975174158211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Bensulide

2 Products
Image of Bensulide
CAS Number:
Empirical Formula (Hill Notation):
 C14H24NO4PS3
Molecular Weight:
 397.51
 Beilstein/REAXYS Number:
 2164989
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Shelf Life
  Special Grade
Show More