• USA Home
  • Search Results

  • bensultap431611760631411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Bensultap

1 Products
Synonym:   S,S′-[2-(Dimethylamino)trimethylene] di(benzenethiosulfonate)
Image of Bensultap
CAS Number:
Empirical Formula (Hill Notation):
 C17H21NO4S4
Molecular Weight:
 431.61
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
Show More