• USA Home
  • Search Results

  • benthiavalicarbisopropyl3814617740668711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Benthiavalicarb isopropyl

1 Products
Image of Benthiavalicarb isopropyl
CAS Number:
Empirical Formula (Hill Notation):
 C18H24FN3O3S
Molecular Weight:
 381.46
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
Show More