• USA Home
  • Search Results

  • bentranil22323102246411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Bentranil

1 Products
Synonym:   2-Phenyl-3,1-benzoxazin-4-one
Image of Bentranil
CAS Number:
Empirical Formula (Hill Notation):
 C14H9NO2
Molecular Weight:
 223.23
 Beilstein/REAXYS Number:
 170624
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
Show More