• USA Home
  • Search Results

  • benzaldehyde1061210052711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Benzaldehyde

11 Products
Synonym:   Bitter almond
Image of Benzaldehyde
CAS Number:
Linear Formula:
 C6H5CHO
Molecular Weight:
 106.12
 Beilstein/REAXYS Number:
 471223
 EC Number:
 MDL number:
 PubChem Substance ID:
 eCl@ss:
 39023701
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Document Links
  Anal. Standard Form
  Allergen
  Organoleptic property
  Purity Grade
  Potency
  Refractive Index
  Special Grade
  Physical Form
  Assay
Show More