• USA Home
  • Search Results

  • benzaldehydednphsolution12345115784211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Benzaldehyde-DNPH solution

1 Products
Synonym:   Benzaldehyde-2,4-DNPH solution, Benzaldehyde-2,4-dinitrophenylhydrazone solution
Image of Benzaldehyde-DNPH solution
CAS Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Titertest
Show More