• USA Home
  • Search Results

  • benzalkoniumchloride10solution123456344941211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Benzalkonium Chloride 10% Solution

1 Products
Synonym:   Alkylbenzyldimethylammonium chloride, Benzalkonium chloride solution
Image of Benzalkonium Chloride 10% Solution
CAS Number:
 MDL number:
 NACRES
 NA.24
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
Show More