• USA Home
  • Search Results

  • benzalkoniumchloride123456344941211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Benzalkonium chloride

3 Products
Synonym:   Alkylbenzyldimethylammonium chloride, Alkyldimethylbenzylammonium chloride
Image of Benzalkonium chloride
CAS Number:
Linear Formula:
 C6H5CH2N(CH3)2RCl (R=C8H17 to C18H37)
 Beilstein/REAXYS Number:
 4062599
 EC Number:
 MDL number:
 NACRES
 NA.22
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Chemical Composition
  Product Line
  ID clarifier
  Agency/Method
Show More