• USA Home
  • Search Results

  • benzalkoniumchloridesolution123456344941211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Benzalkonium chloride solution

2 Products
Synonym:   Alkylbenzyldimethylammonium chloride
Image of Benzalkonium chloride solution
CAS Number:
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Agency/Method
  Physical Form
  Biological Source
  Concentration
Show More