• USA Home
  • Search Results

  • benzbanthracene228299224011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Benz[b]anthracene

2 Products
Synonym:   2,3-Benzanthracene, Naphthacene, Tetracene
Image of Benz[<I>b</I>]anthracene
CAS Number:
Empirical Formula (Hill Notation):
 C18H12
Molecular Weight:
 228.29
 Beilstein/REAXYS Number:
 1909299
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Orbital energy
  OPV Device Performance
  Semiconductor properties
  Special Grade
  Application(s)
  Assay
Show More